missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MOB kinase activator 1A Protein

A DNA sequence encoding the Homo sapiens (Human) MOB kinase activator 1A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8113-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062731

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001304039.1
MOB kinase activator 1A Protein
Research Use Only
MOB1A
Recombinant Protein
E. coli
SFLFSSRSSK TFKPKKNIPE GSHQYELLKH AEATLGSGNL RQAVMLPEGE DLNEWIAVNT VDFFNQINML YGTITEFCTE ASCPVMSAGP RYEYHWADGT NIKKPIKCSA PKYIDYLMTW VQDQLDDETL FPSKIGVPFP KNFMSVAKTI LKRLFRVYAH IYHQHFDSVM QLQEEAHLNT SFKHFIFFVQ EFNLIDRREL APLQELIEKL GSKDR
Greater than 90% as determined by SDS-PAGE.
55233
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers