missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MOBKL3 Protein
A cDNA sequence encoding the MOBKL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12714-2ug
Additional Details : Weight : 0.01000kg
Specifications
MOBKL3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVMAEGTAVL RRNRPGTKAQ DFYNWPDESF DEMDSTLAVQ QYIQQNIRAD CSNIDKILEP PEGQDEGVWK YEHLRQFCLE LNGLAVKLQS ECHPDTCTQM TATEQWIFLC AAHKTPKECP AIDYTRHTLD GAACLLNSNK YFPSRVSIKE SSVAKLGSVC RRIYRIFSHA YFHHRQIFDE YENETFLCHR FTKFVMKYNL MSKDNLIVPI LEEEVQNSVS GESEA | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MOBKL3 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) containing 0.2M Nacl, 5mM DTT, 10% glycerol. |