missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRTO4 Protein
A cDNA sequence encoding the MRTO4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12737-10ug
Additional Details : Weight : 0.01000kg
Specifications
MRTO4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPKSKRD KKVSLTKTAK KGLELKQNLI EELRKCVDTY KYLFIFSVAN MRNSKLKDIR NAWKHSRMFF GKNKVMMVAL GRSPSDEYKD NLHQVSKRLR GEVGLLFTNR TKEEVNEWFT KYTEMDYARA GNKAAFTVSL DPGPLEQFPH SMEPQLRQLG LPTALKRGVV TLLSDYEVCK EGDVLTPEQA RVLKLFGYEM AEFKVTIKYM WDSQSGRFQQ MGDDLPESAS ESTEESDSED DD | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MRTO4 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |