missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MSRB2 Protein

A cDNA sequence encoding the MSRB2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12744-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991399

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

MSRB2 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MSRB2 protein solution (1 mg/1ml) is formulated in In 20 mM Tris-HCl Buffer (pH 7.5), 1 mM DTT, 0.1 mM PMSF and 10% Glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MVRGQAGGGG PGTGPGLGEA GSLATCELPL AKSEWQKKLT PEQFYVTREK GTEPPFSGIY LNNKEAGMYH CVCCDSPLFS SEKKYCSGTG WPSFSEAHGT SGSDESHTGI LRRLDTSLGS ARTEVVCKQC EAHLGHVFPD GPGPNGQRFC INSVALKFKP RKH
Videos
SDS
Documents
Certificates