missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MUCL1 Protein
A cDNA sequence encoding the MUCL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12751-2ug
Additional Details : Weight : 0.01000kg
Specifications
118430 | |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
MUCL1 | |
Human | |
His | |
MUCL1 protein solution (0.25 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol. |
MUCL1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSNPTTAAP ADTYPATGPA DDEAPDAETT AAATTATTAA PTTATTAAST TARKDIPVLP KWVGDLPNGR VCP |