missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mutant GSTO1 Protein
A cDNA sequence encoding the Mutant GSTO1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP14021-HIS-EC-1mg
Specifications
Mutant GSTO1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSGESARSLG KGSAPPGPVP EGSIRIYSMR FCPFAERTRL VLKAKGIRHE VININLKNKP EWFFKKNPFG LVPVLENSQG QLIYESAITC EYLDEAYPGK KLLPDDPYEK ACQKMILELF SKVPSLVGSF IRSQNKEDYA GLKEEFRKEF TKLEEVLTNK KTTFFGGNSI SMIDYLIWPW FERLEAMKLN ECVDHTPKLK LWMAAMKEDP TVSALLTSEK DWQGFLELYL QNSPEACDYG L | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GSTO1 protein is supplied in 1x PBS and 50% glycerol. |