missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYL12B Protein
A cDNA sequence encoding the MYL12B was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | MYL12B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971479
|
enQuireBio™
QP12768-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961479
|
enQuireBio™
QP12768-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951479
|
enQuireBio™
QP12768-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MYL12B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MYL12B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM Nacl, 2mM DTT and 40% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSSKKA KTKTTKKRPQ RATSNVFAMF DQSQIQEFKE AFNMIDQNRD GFIDKEDLHD MLASLGKNPT DAYLDAMMNE APGPINFTMF LTMFGEKLNG TDPEDVIRNA FACFDEEATG TIQEDYLREL LTTMGDRFTD EEVDELYREA PIDKKGNFNY IEFTRILKHG AKDKDD | |
Greater than 90% as determined by SDS-PAGE. |