missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MYL12B Protein

A cDNA sequence encoding the MYL12B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12768-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961479

  • 203.00 € / 20µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MYL12B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSSKKA KTKTTKKRPQ RATSNVFAMF DQSQIQEFKE AFNMIDQNRD GFIDKEDLHD MLASLGKNPT DAYLDAMMNE APGPINFTMF LTMFGEKLNG TDPEDVIRNA FACFDEEATG TIQEDYLREL LTTMGDRFTD EEVDELYREA PIDKKGNFNY IEFTRILKHG AKDKDD
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MYL12B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM Nacl, 2mM DTT and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers