missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myoglobin Protein
A cDNA sequence encoding the Myoglobin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12776-10ug
Additional Details : Weight : 0.01000kg
Specifications
4151 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MGLSDGEWQL VLNVWGKVEA DIPGHGQEVL IRLFKGHPET LEKFDKFKHL KSEDEMKASE DLKKHGATVL TALGGILKKK GHHEAEIKPL AQSHATKHKI PVKYLEFISE CIIQVLQSKH PGDFGADAQG AMNKALELFR KDMASNYKEL GFQG | |
Greater than 96.0%. |
Myoglobin Protein | |
Research Use Only | |
MB | |
Human | |
Untagged | |
The protein solution is in 0.05M phosphate buffer pH 7.5 containing 0.15M NaCl and 0.09% NaN3. Filtered through a 0.2microm membrane. |