missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAGA Protein
A cDNA sequence encoding the NAGA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12785-50ug
Additional Details : Weight : 0.01000kg
Specifications
NAGA Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
LDNGLLQTPP MGWLAWERFR CNINCDEDPK NCISEQLFME MADRMAQDGW RDMGYTYLNI DDCWIGGRDA SGRLMPDPKR FPHGIPFLAD YVHSLGLKLG IYADMGNFTC MGYPGTTLDK VVQDAQTFAE WKVDMLKLDG CFSTPEERAQ GYPKMAAALN ATGRPIAFSC SWPAYEGGLP PRVNYSLLAD ICNLWRNYDD IQDSWWSVLS ILNWFVEHQD ILQPVAGPGH WNDPDMLLIG NFGLSLEQSR AQMALWTVLA APLLMSTDLR TISAQNMDIL QNPLMIKINQ DPLGIQGRRI HKEKSLIEVY MRPLSNKASA LVFFSCRTDM PYRYHSSLGQ LNFTGSVIYE AQDVYSGDII SGLRDETNFT VIINPSGVVM WYLYPIKNLE MSQQHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NAGA protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol. |