missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NBL1 Protein

A cDNA sequence encoding the NBL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12797-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991569

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NBL1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NBL1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSAPPPINK LALFPDKSAW CEAKNITQIV GHSGCEAKSI QNRACLGQCF SYSVPNTFPQ STESLVHCDS CMPAQSMWEI VTLECPGHEE VPRVDKLVEK ILHCSCQACG KEPSHEGLSV YVQGEDGPGS QPGTHPHPHP HPHPGGQTPE PEDPPGAPHT EEEGAED
Videos
SDS
Documents
Certificates