missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDRG1 Protein

A cDNA sequence encoding the NDRG1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10791-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985178

Quantity Price
1 2269.63 € / 1mg
3 2133.68 € / 1mg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NDRG1 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MSREMQDVDL AEVKPLVEKG ETITGLLQEF DVQEQDIETL HGSVHVTLCG TPKGNRPVIL TYHDIGMNHK TCYNPLFNYE DMQEITQHFAVCHVDAPGQQ DGAASFPAGY MYPSMDQLAE MLPGVLQQFG LKSIIGMGTG AGAYILTRFA LNNPEMVEGL VLINVNPCAE GWMDWAASKI SGWTQALPDM VVSHLFGKEE MQSNVEVVHT YRQHIVNDMN PGNLHLFINA YNSRRDLEIE RPMPGTHTVT LQCPALLVVG DSSPAVDAVV ECNSKLDPTK TTLLKMADCG GLPQISQPAK LAEAFKYFVQ GMGYMPSASM TRLMRSRTAS GSSVTSLDGT RSRSHTSEGT RSRSHTSEGT RSRSHTSEGA HLDITPNSGA AGNSAGPKSM EVSCLEHHHH HH
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Measured in a cell proliferation assay using MCF7 cell. The ED50 for this effect is 0.5 - 1.5ng/ml, corresponding to a Specific Activity of 666,000 -2,000,000 IU/mg.
Human
His
The NDRG1 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.1mM PMSF and 10% Glycerol.
Videos
SDS
Documents
Certificates