missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFS4 Protein
A cDNA sequence encoding the NDUFS4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12820-25ug
Additional Details : Weight : 0.01000kg
Specifications
4724 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAQDQTQDTQ LITVDEKLDI TTLTGVPEEH IKTRKVRIFV PARNNMQSGV NNTKKWKMEF DTRERWENPL MGWASTADPL SNMVLTFSTK EDAVSFAEKN GWSYDIEERK VPKPKSKSYG ANFSWNKRTR VSTK | |
Greater than 90.0% as determined by SDS-PAGE. |
NDUFS4 Protein | |
Research Use Only | |
NDUFS4 | |
Human | |
Untagged | |
The NDUFS4 solution contains 20mM Tris pH 8 and 30% glycerol. |