missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neutrophil elastase Protein
A DNA sequence encoding the Homo sapiens (Human) Neutrophil elastase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5978-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001963.1 | |
Neutrophil elastase Protein | |
Research Use Only | |
ELANE | |
Recombinant Protein | |
E. coli | |
IVGGRRARPH AWPFMVSLQL RGGHFCGATL IAPNFVMSAA HCVANVNVRA VRVVLGAHNL SRREPTRQVF AVQRIFENGY DPVNLLNDIV ILQLNGSATI NANVQVAQLP AQGRRLGNGV QCLAMGWGLL GRNRGIASVL QELNVTVVTS LCRRSNVCTL VRGRQAGVCF GDSGSPLVCN GLIHGIASFV RGGCASGLYP DAFAPVAQFV NWIDSIIQRS EDNPCPHPRD PDPASRTH | |
Greater than 90% as determined by SDS-PAGE. |
1991 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |