missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMI Protein
A cDNA sequence encoding the NMI was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12857-1mg
Additional Details : Weight : 0.01000kg
Specifications
NMI Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEADKDDTQQ ILKEHSPDEF IKDEQNKGLI DEITKKNIQLK KEIQKLETE LQEATKEFQI KEDIPETKMK FLSVETPEND SQLSNISCSF QVSSKVPYEI QKGQALITFE KEEVAQNVVS MSKHHVQIKDV NLEVTAKPV PLNSGVRFQV YVEVSKMKIN VTEIPDTLRE DQMRDKLELS FSKSRNGGGE VDRVDYDRQS GSAVITFVEI GVADKILKKKEYPLYINQTC HRVTVSPYTE IHLKKYQIFS GTSKRTVLLT GMEGIQMDEE IVEDLINIHF QRAKNGGGEV DVVKCSLGQP HIAYFEE | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NMI protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)1mM DTT and 20% glycerol. |