missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMT1 Protein
A cDNA sequence encoding the NMT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12860-20ug
Additional Details : Weight : 0.01000kg
Specifications
NMT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADESET AVKPPAPPLP QMMEGNGNGH EHCSDCENEE DNSYNRGGLS PANDTGAKKK KKKQKKKKEK GSETDSAQDQ PVKMNSLPAE RIQEIQKAIE LFSVGQGPAK TMEEASKRSY QFWDTQPVPK LGEVVNTHGP VEPDKDNIRQ EPYTLPQGFT WDALDLGDRG VLKELYTLLN ENYVEDDDNM FRFDYSPEFL LWALRPPGWL PQWHCGVRVV SSRKLVGFIS AIPANIHIYD TEKKMVEINF LCVHKKLRSK RVAPVLIREI TRRVHLEGIF QAVYTAGVVL PKPVGTCRYW HRSLNPRKLI EVKFSHLSRN MTMQRTMKLY RLPETPKTAG LRPMETKDIP VVHQLLTRYL KQFHLTPVMS QEEVEHWFYP QENIIDTFVV ENANGEVTDF LSFYTLPSTI MNHPTHKSLK AAYSFYNVHT QTPLLDLMSD ALVLAKMKGF DVFNALDLME NKTFLEKLKF GIGDGNLQYY LYNWKCPSMG AEKVGLVLQ | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NMT1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4). |