missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NT5M Protein
A cDNA sequence encoding the NT5M was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12890-50ug
Additional Details : Weight : 0.01000kg
Specifications
NT5M Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGGRALRVLV DMDGVLADFE GGFLRKFRAR FPDQPFIALE DRRGFWVSEQ YGRLRPGLSE KAISIWESKN FFFELEPLPG AVEAVKEMAS LQNTDVFICT SPIKMFKYCP YEKYAWVEKY FGPDFLEQIV LTRDKTVVSA DLLIDDRPDI TGAEPTPSWE HVLFTACHNQ HLQLQPPRRR LHSWADDWKA ILDSKRPC | |
Greater than 95% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NT5M protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |