missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUBP2 Protein
A cDNA sequence encoding the NUBP2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | NUBP2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951879
|
enQuireBio™
QP12895-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931879
|
enQuireBio™
QP12895-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941879
|
enQuireBio™
QP12895-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NUBP2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUBP2 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 50% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEAAAEP GNLAGVRHII LVLSGKGGVG KSTISTELAL ALRHAGKKVG ILDVDLCGPS IPRMLGAQGR AVHQCDRGWA PVFLDREQSI SLMSVGFLLE KPDEAVVWRG PKKNALIKQF VSDVAWGELD YLVVDTPPGT SDEHMATIEA LRPYQPLGAL VVTTPQAVSV GDVRRELTFC RKTGLRVMGI VENMSGFTCP HCTECTSVFS RGGGEELAQL AGVPFLGSVP LDPALMRTLE EGHDFIQEFP GSPAFAALTS IAQKILDATP ACLP | |
Greater than 90.0% as determined by SDS-PAGE. |