missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NUDT5 Protein

A cDNA sequence encoding the NUDT5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12907-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991909

  • 3401.86 € / 1mg

Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NUDT5 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NUDT5 Human 1 mg/ml solution containing 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MESQEPTESS QNGKQYIISE ELISEGKWVK LEKTTYMDPT GKTRTWESVK RTTRKEQTAD GVAVIPVLQR TLHYECIVLV KQFRPPMGGY CIEFPAGLID DGETPEAAAL RELEEETGYK GDIAECSPAV CMDPGLSNCT IHIVTVTING DDAENARPKP KPGDGEFVEV ISLPKNDLLQ RLDALVAEEH LTVDARVYSY ALALKHANAK PFEVPFLKF
Videos
SDS
Documents
Certificates