missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUP62 Protein
A cDNA sequence encoding the NUP62 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5655.00 €
Specifications
Name | NUP62 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971919
|
enQuireBio™
QP12909-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961919
|
enQuireBio™
QP12909-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951919
|
enQuireBio™
QP12909-1MG |
1 mg |
5655.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NUP62 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUP62 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MSGFNFGGTG APTGGFTFGT AKTATTTPAT GFSFSTSGTG GFNFGAPFQP ATSTPSTGLF SLATQTPATQ TTGFTFGTAT LASGGTGFSL GIGASKLNLS NTAATPAMAN PSGFGLGSSN LTNAISSTVT SSQGTAPTGF VFGPSTTSVA PATTSGGFSF TGGSTAQPSG FNIGSAGNSA QPTAPATLPF TPATPAATTA GATQPAAPTP TATITSTGPS LFASIATAPT SSATTGLSLC TPVTTAGAPT AGTQGFSLKA PGAASGTSTT TSTAATATAT TTSSSSTTGF ALNLKPLAPA GIPSNTAAAV TAPPGPGAAA GAAASSAMTY AQLESLINKW SLELEDQERH FLQQATQVNA WDRTLIENGE KITSLHREVE KVKLDQKRLD QELDFILSQQ KELEDLLSPL EELVKEQSGT IYLQHADEER EKTYKLAENI DAQLKRMAQD LKDIIEHLNT SGAPADTSDP LQQICKILNA HMDSLQWIDQ NSALLQRKVE EVTKVCEGRR KEQERSFRIT FD | |
Greater than 80% as determined by SDS-PAGE. |