missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUP62CL Protein
A cDNA sequence encoding the NUP62CL was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12910-20ug
Additional Details : Weight : 0.01000kg
Specifications
NUP62CL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQFTSIS NSLTSTAAIG LSFTTSTTTT ATFTTNTTTT ITSGFTVNQN QLLSRGFENL VPYTSTVRFV FYMEK | |
Greater than 80.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUP62CL protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |