missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OAZ1 Protein

A cDNA sequence encoding the OAZ1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12916-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991939

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

OAZ1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The OAZ1 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMVKSSLQ RILNSHCFAR EKEGDKPSAT IHASRTMPLL SLHSRGGSSS ESSRVSLHCC SNPGPGPRWC SDAPHPPLKI PGGRGNSQRD HNLSANLFYS DDRLNVTEEL TSNDKTRILN VQSRLTDAKR INWRTVLSGG SLYIEIPGGA LPEGSKDSFA VLLEFAEEQL RADHVFICFH KNREDRAALL RTFSFLGFEI VRPGHPLVPK RPDACFMAYT FERESSGEEE E
Videos
SDS
Documents
Certificates