missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OCT Protein
A cDNA sequence encoding the OCT was constructed and used to recombinantly synthesize the protein.
131.00 € - 1305.00 €
Specifications
Name | OCT Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15911959
|
enQuireBio™
QP12920-1MG |
1 mg |
131.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931959
|
enQuireBio™
QP12920-5MG |
5 mg |
324.00 €
5mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921959
|
enQuireBio™
QP12920-25MG |
25 mg |
1305.00 €
25mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
OCT Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The Octreotide was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
H-D-Phe-Cys-Phe-D-Trp-Lys-Thr-Cys-L-threoninol | |
Greater than 98.0% as determined by RP-HPLC. |