missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Orofacial cleft 1 candidate gene 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Orofacial cleft 1 candidate gene 1 protein, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Name | Orofacial cleft 1 candidate gene 1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | OFCC1 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030921
|
enQuireBio™
QP7807-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050921
|
enQuireBio™
QP7807-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040921
|
enQuireBio™
QP7807-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060921
|
enQuireBio™
QP7807-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070921
|
enQuireBio™
QP7807-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080921
|
enQuireBio™
QP7807-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Orofacial cleft 1 candidate gene 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
OFCC1 | |
Recombinant Protein | |
E. coli | |
REKFQQKALK QTKQKKSKSA EFLMVKEDRE ATEGTGNPAF NMSSPDLSAC QTAEKKVIRH DMPDRTLAAH QQKFRLPASA EPKGNEYGRN YFDPLMDEEI NPRQCATEVS REDDDRIFYN RLTKLFDESR QGEPQDESGR EETLNSEAPG SSNKSHEIHK EASEATTAHL EEFQRSQKTI ILLGSSPLEQ EIRSTSLHCM EDEMSHPWIL LLKVTAVIRS RRYYREQRF | |
Greater than 90% as determined by SDS-PAGE. |