missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human p53 / TP53 Protein

A DNA sequence encoding the Homo sapiens (Human) p53 / TP53, was expressed in the hosts and tags indicated.

160.00 € - 2285.00 €

Specifications

Accession Number NP_000537.3
Gene ID (Entrez) 7157
Name p53 / TP53 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16036080
View Documents Promotion Details
enQuireBio™
QP6822-EC-100UG
100 μg
505.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16056080
View Documents Promotion Details
enQuireBio™
QP6822-EC-1MG
1 mg
2285.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16046080
View Documents Promotion Details
enQuireBio™
QP6822-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16066080
View Documents Promotion Details
enQuireBio™
QP6822-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16076080
View Documents Promotion Details
enQuireBio™
QP6822-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16086080
View Documents Promotion Details
enQuireBio™
QP6822-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_000537.3
p53 / TP53 Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
7157
Research Use Only
TP53
Recombinant Protein
E. coli
MEEPQSDPSV EPPLSQETFS DLWKLLPENN VLSPLPSQAM DDLMLSPDDI EQWFTEDPGP DEAPRMPEAA PPVAPAPAAP TPAAPAPAPS WPLSSSVPSQ KTYQGSYGFR LGFLHSGTAK SVTCTYSPAL NKMFCQLAKT CPVQLWVDST PPPGTRVRAM AIYKQSQHMT EVVRRCPHHE RCSDSDGLAP PQHLIRVEGN LRVEYLDDRN TFRHSVVVPY EPPEVGSDCT TIHYNYMCNS SCMGGMNRRP ILTIITLEDS SGNLLGRNSF EVRVCACPGR DRRTEEENLR KKGEPHHELP PGSTKRALPN NTSSSPQPKK KPLDGEYFTL QIRGRERFEM FRELNEALEL KDAQAGKEPG GSRAHSSHLK SKKGQSTSRH KKLMFKTEGP DSD
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications