missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAPSS1 Protein
A cDNA sequence encoding the PAPSS1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | PAPSS1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15902079
|
enQuireBio™
QP12958-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15992069
|
enQuireBio™
QP12958-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982069
|
enQuireBio™
QP12958-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PAPSS1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PAPSS1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMRATNV TYQAHHVSRN KRGQVVGTRG GFRGCTVWLT GLSGAGKTTV SMALEEYLVC HGIPCYTLDG DNIRQGLNKN LGFSPEDREE NVRRIAEVAK LFADAGLVCI TSFISPYTQD RNNARQIHEG ASLPFFEVFV DAPLHVCEQR DVKGLYKKAR AGEIKGFTGI DSEYEKPEAP ELVLKTDSCD VNDCVQQVVE LLQERDIVPV DASYEVKELY VPENKLHLAK TDAETLPALK INKVDMQWVQ VLAEGWATPL NGFMREREYL QCLHFDCLLD GGVINLSVPI VLTATHEDKE RLDGCTAFAL MYEGRRVAIL RNPEFFEHRK EERCARQWGT TCKNHPYIKM VMEQGDWLIG GDLQVLDRVY WNDGLDQYRL TPTELKQKFK DMNADAVFAF QLRNPVHNGH ALLMQDTHKQ LLERGYRRPV LLLHPLGGWT KDDDVPLMWR MKQHAAVLEE GVLNPETTVV AIFPSPMMYA GPTEVQWHCR ARMVAGANFY IVGRDPAGMP HPETGKDLYE PSHGAKVLTM APGLITLEIV PFRVAAYNKK KKRMDYYDSE HHEDFEFISG TRMRKLAREG QKPPEGFMAP KAWTVLTEYY KSLEKA | |
Greater than 85% as determined by SDS-PAGE. |