missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Paralemmin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Paralemmin-1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001035224.1 |
---|---|
Gene ID (Entrez) | 5064 |
Name | Paralemmin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043490
|
enQuireBio™
QP6465-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063490
|
enQuireBio™
QP6465-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053490
|
enQuireBio™
QP6465-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073490
|
enQuireBio™
QP6465-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083490
|
enQuireBio™
QP6465-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093490
|
enQuireBio™
QP6465-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001035224.1 | |
Paralemmin-1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5064 | |
Research Use Only | |
PALM | |
Recombinant Protein | |
E. coli | |
MEVLAAETTS QQERLQAIAE KRKRQAEIEN KRRQLEDERR QLQHLKSKAL RERWLLEGTP SSASEGDEDL RRQMQDDEQK TRLLEDSVSR LEKEIEVLER GDSAPATAKE NAAAPSPVRA PAPSPAKEER KTEVVMNSQQ TPVGTPKDKR VSNTPLRTVD GSPMMKAAMY SVEITVEKDK VTGETRVLSS TTLLPRQPLP LGIKVYEDET KVVHAVDGTA ENGIHPLSSS EVDELIHKAD EVTLSEAGST AGAAETRGAV EGAARTTPSR REITGVQAQP GEATSGPPGI QPGQEPPVTM IFMGYQNVED EAETKKVLGL QDTITAELVV IEDAAEPKEP APPNGSAAEP PTEAASREEN QAGPEATTSD PQDLDMKKHR CKCC | |
Greater than 90% as determined by SDS-PAGE. |