missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Parathyroid hormone 2 receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Parathyroid hormone 2 receptor, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001296445.1 |
---|---|
Gene ID (Entrez) | 5746 |
Name | Parathyroid hormone 2 receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034100
|
enQuireBio™
QP6552-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054100
|
enQuireBio™
QP6552-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044100
|
enQuireBio™
QP6552-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064100
|
enQuireBio™
QP6552-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074100
|
enQuireBio™
QP6552-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084100
|
enQuireBio™
QP6552-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001296445.1 | |
Parathyroid hormone 2 receptor Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5746 | |
Research Use Only | |
PTH2R | |
Recombinant Protein | |
E. coli | |
DSDGTITIEE QIVLVLKAKV QCELNITAQL QEGEGNCFPE WDGLICWPRG TVGKISAVPC PPYIYDFNHK GVAFRHCNPN GTWDFMHSLN KTWANYSDCL RFLQPDISIG KQEFFERLY | |
Greater than 90% as determined by SDS-PAGE. |