missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAX8 Protein
A cDNA sequence encoding the PAX8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12969-2ug
Additional Details : Weight : 0.01000kg
Specifications
PAX8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPHNSIR SGHGGLNQLG GAFVNGRPLP EVVRQRIVDL AHQGVRPCDI SRQLRVSHGC VSKILGRYYE TGSIRPGVIG GSKPKVATPK VVEKIGDYKR QNPTMFAWEI RDRLLAEGVC DNDTVPSVSS INRIIRTKVQ QPFNLPMDSC VATKSLSPGH TLIPSSAVTP PESPQSDSLG STYSINGLLG IAQPGSDKRK MDDSDQDSCR LSIDSQSSSS GPRKHLRTDA FSQHHLEPLE CPFERQHYPE AYASPSHTKG EQEVNTLAMP MATPPTPPTA RPGASPTPAC | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PAX8 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |