missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZK1 Protein
A DNA sequence encoding the Homo sapiens (Human) PDZK1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001188254.1 |
---|---|
Gene ID (Entrez) | 5174 |
Name | PDZK1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16020311
|
enQuireBio™
QP7705-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040311
|
enQuireBio™
QP7705-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030311
|
enQuireBio™
QP7705-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050311
|
enQuireBio™
QP7705-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060311
|
enQuireBio™
QP7705-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070311
|
enQuireBio™
QP7705-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001188254.1 | |
PDZK1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5174 | |
Research Use Only | |
PDZK1 | |
Recombinant Protein | |
E. coli | |
MTSTFNPREC KLSKQEGQNY GFFLRIEKDT EGHLVRVVEK CSPAEKAGLQ DGDRVLRING VFVDKEEHMQ VVDLVRKSGN SVTLLVLDGD SYEKAVKTRV DLKELGQSQK EQGLSDNILS PVMNGGVQTW TQPRLCYLVK EGGSYGFSLK TVQGKKGVYM TDITPQGVAM RAGVLADDHL IEVNGENVED ASHEEVVEKV KKSGSRVMFL LVDKETDKRH VEQKIQFKRE TASLKLLPHQ PRIVEMKKGS NGYGFYLRAG SEQKGQIIKD IDSGSPAEEA GLKNNDLVVA VNGESVETLD HDSVVEMIRK GGDQTSLLVV DKETDNMYRL AHFSPFLYYQ SQELPNGSVK EAPAPTPTSL EVSSPPDTTE EVDHKPKLCR LAKGENGYGF HLNAIRGLPG SFIKEVQKGG PADLAGLEDE DVIIEVNGVN VLDEPYEKVV DRIQSSGKNV TLLVCGKKAY DYFQAKKIPI VSSLADPLDT PPDSKEGIVV ESNHDSHMAK ERAHSTASHS SSNSEDTEM | |
Greater than 90% as determined by SDS-PAGE. |