missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peptidyl-prolyl cis-trans isomerase-like 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Peptidyl-prolyl cis-trans isomerase-like 4, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_624311.1 |
---|---|
Gene ID (Entrez) | 85313 |
Name | Peptidyl-prolyl cis-trans isomerase-like 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031941
|
enQuireBio™
QP7976-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051941
|
enQuireBio™
QP7976-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041941
|
enQuireBio™
QP7976-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061941
|
enQuireBio™
QP7976-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071941
|
enQuireBio™
QP7976-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081941
|
enQuireBio™
QP7976-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_624311.1 | |
Peptidyl-prolyl cis-trans isomerase-like 4 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
85313 | |
Research Use Only | |
PPIL4 | |
Recombinant Protein | |
E. coli | |
MAVLLETTLG DVVIDLYTEE RPRACLNFLK LCKIKYYNYC LIHNVQRDFI IQTGDPTGTG RGGESIFGQL YGDQASFFEA EKVPRIKHKK KGTVSMVNNG SDQHGSQFLI TTGENLDYLD GVHTVFGEVT EGMDIIKKIN ETFVDKDFVP YQDIRINHTV ILDDPFDDPP DLLIPDRSPE PTREQLDSGR IGADEEIDDF KGRSAEEVEE IKAEKEAKTQ AILLEMVGDL PDADIKPPEN VLFVCKLNPV TTDEDLEIIF SRFGPIRSCE VIRDWKTGES LCYAFIEFEK EEDCEKAFFK MDNVLIDDRR IHVDFSQSVA KVKWKGKGGK YTKSDFKEYE KEQDKPPNLV LKDKVKPKQD TKYDLILDEQ AEDSKSSHSH TSKKHKKKTH HCSEEKEDED YMPIKNTNQD IYREMGFGHY EEEESCWEKQ KSEKRDRTQN RSRSRSRERD GHYSNSHKSK YQTDLYERER SKKRDRSRSP KKSKDKEKSK YR | |
Greater than 90% as determined by SDS-PAGE. |