missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PFDN1 Protein
A DNA sequence encoding the Homo sapiens (Human) PFDN1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8371-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002613.2 | |
PFDN1 Protein | |
Research Use Only | |
PFDN1 | |
Recombinant Protein | |
E. coli | |
AAPVDLELKK AFTELQAKVI DTQQKVKLAD IQIEQLNRTK KHAHLTDTEI MTLVDETNMY EGVGRMFILQ SKEAIHSQLL EKQKIAEEKI KELEQKKSYL ERSVKEAEDN IREMLMARRA Q | |
Greater than 90% as determined by SDS-PAGE. |
5201 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |