missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGDH / PHGDH Protein
A DNA sequence encoding the Homo sapiens (Human) PGDH / PHGDH, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_006614.2 |
---|---|
Gene ID (Entrez) | 26227 |
Name | PGDH / PHGDH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024621
|
enQuireBio™
QP8438-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044621
|
enQuireBio™
QP8438-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034621
|
enQuireBio™
QP8438-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054621
|
enQuireBio™
QP8438-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064621
|
enQuireBio™
QP8438-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074621
|
enQuireBio™
QP8438-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006614.2 | |
PGDH / PHGDH Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
26227 | |
Research Use Only | |
PHGDH | |
Recombinant Protein | |
E. coli | |
AFANLRKVLI SDSLDPCCRK ILQDGGLQVV EKQNLSKEEL IAELQDCEGL IVRSATKVTA DVINAAEKLQ VVGRAGTGVD NVDLEAATRK GILVMNTPNG NSLSAAELTC GMIMCLARQI PQATASMKDG KWERKKFMGT ELNGKTLGIL GLGRIGREVA TRMQSFGMKT IGYDPIISPE VSASFGVQQL PLEEIWPLCD FITVHTPLLP STTGLLNDNT FAQCKKGVRV VNCARGGIVD EGALLRALQS | |
Greater than 90% as determined by SDS-PAGE. |