missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PHD finger-like domain-containing protein 5A Protein
A DNA sequence encoding the Homo sapiens (Human) PHD finger-like domain-containing protein 5A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6490-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_116147.1 | |
PHD finger-like domain-containing protein 5A Protein | |
Research Use Only | |
PHF5A | |
Recombinant Protein | |
E. coli | |
MAKHHPDLIF CRKQAGVAIG RLCEKCDGKC VICDSYVRPC TLVRICDECN YGSYQGRCVI CGGPGVSDAY YCKECTIQEK DRDGCPKIVN LGSSKTDLFY ERKKYGFKKR | |
Greater than 90% as determined by SDS-PAGE. |
84844 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |