missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PHF13 Protein
A cDNA sequence encoding the PHF13 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1595.00 €
Specifications
Name | PHF13 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15922319
|
enQuireBio™
QP13038-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942319
|
enQuireBio™
QP13038-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932319
|
enQuireBio™
QP13038-50UG |
50 μg |
1595.00 €
50µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PHF13 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PHF13 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT, 1mM EDTA and 250mM Imidazole. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDSDSCA AAFHPEEYSP SCKRRRTVED FNKFCTFVLA YAGYIPYPKE ELPLRSSPSP ANSTAGTIDS DGWDAGFSDI ASSVPLPVSD RCFSHLQPTL LQRAKPSNFL LDRKKTDKLK KKKKRKRRDS DAPGKEGYRG GLLKLEAADP YVETPTSPTL QDIPQAPSDP CSGWDSDTPS SGSCATVSPD QVKEIKTEGK RTIVRQGKQV VFRDEDSTGN DEDIMVDSDD DSWDLVTCFC MKPFAGRPMI ECNECHTWIH LSCAKIRKSN VPEVFVCQKC RDSKFDIRRS NRSRTGSRKL FLD | |
Greater than 85% as determined by SDS-PAGE. |