missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PI3 / Elafin / WFDC14 Protein
A DNA sequence encoding the Homo sapiens (Human) PI3 / Elafin / WFDC14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6493-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002629.1 | |
PI3 / Elafin / WFDC14 Protein | |
Research Use Only | |
PI3 | |
Recombinant Protein | |
Yeast | |
AQEPVKGPVS TKPGSCPIIL IRCAMLNPPN RCLKDTDCPG IKKCCEGSCG MACFVPQ | |
Greater than 90% as determined by SDS-PAGE. |
5266 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |