missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PI3Kb Protein
A cDNA sequence encoding the PI3Kb was constructed and used to recombinantly synthesize the protein.
203.00 € - 665.00 €
Specifications
Name | PI3Kb Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The specific activity was found to be 3,000 units/mg (1 unit is defined as 1 picomole phosphate transferred to PIP2 per minute). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985258
|
enQuireBio™
QP10818-1UG |
1 μg |
203.00 €
1µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995258
|
enQuireBio™
QP10818-2.5UG |
2.5 μg |
373.00 €
2.50µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905268
|
enQuireBio™
QP10818-5UG |
5 μg |
665.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PI3Kb Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
0.9 mg/ml solution in PBS and 2mM MgCl2. |
Research Use Only | |
The specific activity was found to be 3,000 units/mg (1 unit is defined as 1 picomole phosphate transferred to PIP2 per minute). | |
Human | |
Untagged | |
Greater than 90.0% as determined by SDS Page. |