missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PIN4 Protein

A cDNA sequence encoding the PIN4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10821-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985268

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

5303
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
PIN4
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPMAGLLKGL VRQLERFSVQ QQASKMPPKG KSGSGKAGKG GAASGSDSAD KKAQGPKGGG NAVKVRHILC EKHGKIMEAM EKLKSGMRFN EVAAQYSEDK ARQGGDLGWM TRGSMVGPFQ EAAFALPVSG MDKPVFTDPP VKTKFGYHII MVEGRK
PIN4 Protein
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 300 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-HCl pH8.0 using chymotrypsin.
Human
His
PIN4 solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1mM PMSF.
Videos
SDS
Documents
Certificates