missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIP4K2B Protein
A cDNA sequence encoding the PIP4K2B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13058-1mg
Specifications
PIP4K2B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSNCTS TTAVAVAPLS ASKTKTKKKH FVCQKVKLFR ASEPILSVLM WGVNHTINEL SNVPVPVMLM PDDFKAYSKI KVDNHLFNKE NLPSRFKFKE YCPMVFRNLR ERFGIDDQDY QNSVTRSAPI NSDSQGRCGT RFLTTYDRRF VIKTVSSEDV AEMHNILKKY HQFIVECHGN TLLPQFLGMY RLTVDGVETY MVVTRNVFSH RLTVHRKYDL KGSTVAREAS DKEKAKDLPT FKDNDFLNEG QKLHVGEESK KNFLEKLKRD VEFLAQLKIM DYSLLVGIHD VDRAEQEEME VEERAEDEEC ENDGVGGNLL CSYGTPPDSP GNLLSFPRFF GPGEFDPSVD VYAMKSHESS PKKEVYFMAI IDILTPYDTK KKAAHAAKTV KHGAGAEIST VNPEQYSKRF NEFMSNILT | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PIP4K2B protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |