missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Plexin-A4 Protein
A DNA sequence encoding the Homo sapiens (Human) Plexin-A4, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001099013.1 |
---|---|
Gene ID (Entrez) | 91584 |
Name | Plexin-A4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042411
|
enQuireBio™
QP8056-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062411
|
enQuireBio™
QP8056-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052411
|
enQuireBio™
QP8056-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072411
|
enQuireBio™
QP8056-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082411
|
enQuireBio™
QP8056-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092411
|
enQuireBio™
QP8056-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001099013.1 | |
Plexin-A4 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
91584 | |
Research Use Only | |
PLXNA4 | |
Recombinant Protein | |
E. coli | |
TLLTRQPAPL SQKQRSFVTF RGEPAEGFNH LVVDERTGHI YLGAVNRIYK LSSDLKVLVT HETGPDEDNP KCYPPRIVQT CNEPLTTTNN VNKMLLIDYK ENRLIACGSL YQGICKLLRL EDLFKLGEPY HKKEHYLSGV NESGSVFGVI VSYSNLDDKL FIATAVDGKP EYFPTISSRK LTKNSEADGM FAYVFHDEFV ASMIKIPSDT FTIIPDFDIY YVYGFSSGNF VYFLTLQPEM VSPPGSTTKE QVYTSKLVRL CKEDTAFNSY VEVPIGCERS GVEYRLLQAA YLSKAGAVLG RTLGVHPDDD LLFTVFSKGQ KRKMKSLDES ALCIFILKQI NDRIKERLQS CYRGEGTLDL AWLKVKDIPC SSALLTIDDN FCGLDMNAPL GVSDMVRGIP VFTEDRDRMT SVIAYVYKNH SLAFVGTKSG KLKKIRVDGP RGNALQYETV QVVDPGPVLR DMAFSKDHEQ LYIMSERQLT RVPVESCGQY QSCGECLGS | |
Greater than 90% as determined by SDS-PAGE. |