missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PON2 Protein
A cDNA sequence encoding the PON2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6960.00 €
Specifications
Name | PON2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932539
|
enQuireBio™
QP13110-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912539
|
enQuireBio™
QP13110-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922539
|
enQuireBio™
QP13110-1MG |
1 mg |
6960.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PON2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PON2 is supplied in PBS and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGRLVAVGLLGIALALLGERLLALRNRLKASREVESVDLPHCHLIKGIEAGSEDID ILPNGLAFFSVGLKFPGLHSFAPDKPGGILMMDLKEEKPRARELRISRGFDLASFNP HGISTFIDNDDTVYLFVVNHPEFKNTVEIFKFEEAENSLLHLKTVKHELLPSVNDIT AVGPAHFYATNDHYFSDPFLKYLETYLNLHWANVVYYSPNEVKVVAEGFDSAN GINISPDDKYIYVADILAHEIHVLEKHTNMNLTQLKVLELDTLVDNLSIDPSSGDIW VGCHPNGQKLFVYDPNNPPSSEVLRIQNILSEKPTVTTVYANNGSVLQGSSVASVY DGKLLIGTLYHRALYCELZ | |
Greater than 95% as determined by SDS-PAGE. Single band on Western Blot. |