missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPIAL4A Protein
A DNA sequence encoding the Homo sapiens (Human) PPIAL4A, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001137355.1 |
---|---|
Gene ID (Entrez) | 653505 |
Name | PPIAL4A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013671
|
enQuireBio™
QP8269-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033671
|
enQuireBio™
QP8269-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023671
|
enQuireBio™
QP8269-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043671
|
enQuireBio™
QP8269-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053671
|
enQuireBio™
QP8269-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063671
|
enQuireBio™
QP8269-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001137355.1 | |
PPIAL4A Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
653505 | |
Research Use Only | |
PPIAL4A | |
Recombinant Protein | |
E. coli | |
MVNSVVFFDI TVDGKPLGRI SIKLFADKIL KTAENFRALS TGEKGFRYKG SCFHRIIPGF MCQGGDFTRH NGTGDKSIYG EKFDDENLIR KHTGSGILSM ANAGPNTNGS QFFICAAKTE WLDGKHVAFG KVKERVNIVE AMEHFGYRNS KTSKKITIAD CGQF | |
Greater than 90% as determined by SDS-PAGE. |