missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1R1A Protein
A cDNA sequence encoding the PPP1R1A was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | PPP1R1A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932579
|
enQuireBio™
QP13124-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922579
|
enQuireBio™
QP13124-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912579
|
enQuireBio™
QP13124-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PPP1R1A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PPP1R1A solution contains 50mM Tris pH 8.0, 0.1mM PMSF, 1mM EDTA, 1mM DTT and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEQDNSPRKI QFTVPLLEPH LDPEAAEQIR RRRPTPATLV LTSDQSSPEI DEDRIPNPHL KSTLAMSPRQ RKKMTRITPT MKELQMMVEH HLGQQQQGEE PEGAAESTGT QESRPPGIPD TEVESRLGTS GTAKKTAECI PKTHERGSKE PSTKEPSTHI PPLDSKGANS VLEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |