missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRDX2 Protein

A cDNA sequence encoding the PRDX2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10838-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985318

  • 4534.09 € / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PRDX2 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MASGNARIGK PAPDFKATAV VDGAFKEVKL SDYKGKYVVL FFYPLDFTFV CPTEIIAFSN RAEDFRKLGC EVLGVSVDSQ FTHLAWINTP RKEGGLGPLN IPLLADVTRR LSEDYGVLKT DEGIAYRGLF IIDGKGVLRQ ITVNDLPVGR SVDEALRLVQ AFQYTDEHGE VCPAGWKPGS DTIKPNVDDS KEYFSKHN
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
The specific activity was found to be approximately 200-230 pmole/min/microg. The enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX2 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for 1 minute.
Human
Untagged
The Peroxiredoxin solution contains 20mM Tris-HCl pH 8.0, and 10% glycerol.
Videos
SDS
Documents
Certificates