missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable dimethyladenosine transferase Protein
A DNA sequence encoding the Homo sapiens (Human) Probable dimethyladenosine transferase, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001335005.1 |
---|---|
Gene ID (Entrez) | 27292 |
Name | Probable dimethyladenosine transferase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043581
|
enQuireBio™
QP8253-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063581
|
enQuireBio™
QP8253-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053581
|
enQuireBio™
QP8253-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073581
|
enQuireBio™
QP8253-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083581
|
enQuireBio™
QP8253-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093581
|
enQuireBio™
QP8253-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001335005.1 | |
Probable dimethyladenosine transferase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
27292 | |
Research Use Only | |
DIMT1 | |
Recombinant Protein | |
E. coli | |
MPKVKSGAIG RRRGRQEQRR ELKSAGGLMF NTGIGQHILK NPLIINSIID KAALRPTDVV LEVGPGTGNM TVKLLEKAKK VVACELDPRL VAELHKRVQG TPVASKLQVL VGDVLKTDLP FFDTCVANLP YQISSPFVFK LLLHRPFFRC AILMFQREFA LRLVAKPGDK LYCRLSINTQ LLARVDHLMK VGKNNFRPPP KVESSVVRIE PKNPPPPINF QEWDGLVRIT FVRKNKTLSA AFKSSAVQQL LEKNYRIHCS VHNIIIPEDF SIADKIQQIL TSTGFSDKRA RSMDIDDFIR LLHGFNAEGI HFS | |
Greater than 90% as determined by SDS-PAGE. |