missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable histidine--tRNA ligase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Probable histidine--tRNA ligase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6143-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001265660.1 | |
Probable histidine--tRNA ligase, mitochondrial Protein | |
Research Use Only | |
HARS2 | |
Recombinant Protein | |
E. coli | |
QSQVAEAVLT SQLKAHQEKP NFIIKTPKGT RDLSPQHMVV REKILDLVIS CFKRHGAKGM DTPAFELKET LTEKYGEDSG LMYDLKDQGG ELLSLRYDLT VPFARYLAMN KVKKMKRYHV GKVWRRESPT IVQGRYREFC QCDFDIAGQF DPMIPDAECL KIMCEILSGL QLGDFLIKVN DRRIVDGMFA VCGVPESKFR AICSSIDKLD KMAWKDVRHE MVVKKGLAPE VADRIGDYVQ CHGGVSLVEQ MFQDPRLSQN KQALEGLGDL KLLFEYLTLF GIADKISFDL SLARGLDYYT GVIYEAVLLQ TPTQAGEEPL NVGSVAAGGR YDGLVGMFDP KGHKVPCVGL SIGVERIFYI VEQRMKTKGE KVRTTETQVF VATPQKNFLQ ERLKLIAELW DSGIKAEMLY KNNPKLLTQL HYCESTGIPL VVIIGEQELK EGVIKIRSVA SREEVAIKRE NFVAEIQKRL SES | |
Greater than 90% as determined by SDS-PAGE. |
23438 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |