missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prolactin / PRL Protein
Recombinant Protein
Brand: enQuireBio™ QP5359-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000939.1 | |
Prolactin / PRL Protein | |
Research Use Only | |
PRL | |
Recombinant Protein | |
E. coli | |
MLPICPGGAA RCQVTLRDLF DRAVVLSHYI HNLSSEMFSE FDKRYTHGRG FITKAINSCH TSSLATPEDK EQAQQMNQKD FLSLIVSILR SWNEPLYHLV TEVRGMQEAP EAILSKAVEI EEQTKRLLEG MELIVSQVHP ETKENEIYPV WSGLPSLQMA DEESRLSAYY NLLHCLRRDS HKIDNYLKLL KCRIIHNNNC | |
Greater than 95% as determined by SDS-PAGE. |
5617 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the NB2-11 cell proliferation assay. The bioactivity acceptance criteria is set at 1.0 x 10^6 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |