missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prostate stem cell antigen Protein
A DNA sequence encoding the Homo sapiens (Human) Prostate stem cell antigen, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Name | Prostate stem cell antigen Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | PSCA |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16006211
|
enQuireBio™
QP8733-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046211
|
enQuireBio™
QP8733-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016211
|
enQuireBio™
QP8733-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036211
|
enQuireBio™
QP8733-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056211
|
enQuireBio™
QP8733-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026211
|
enQuireBio™
QP8733-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Prostate stem cell antigen Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
PSCA | |
Recombinant Protein | |
Yeast | |
LLCYSCKAQV SNEDCLQVEN CTQLGEQCWT ARIRAVGLLT VISKGCSLNC VDDSQDYYVG KKNITCCDTD LCNAS | |
Greater than 90% as determined by SDS-PAGE. |