missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein quaking Protein
A DNA sequence encoding the Homo sapiens (Human) Protein quaking, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001288014.1 |
---|---|
Gene ID (Entrez) | 9444 |
Name | Protein quaking Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031581
|
enQuireBio™
QP7918-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051581
|
enQuireBio™
QP7918-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041581
|
enQuireBio™
QP7918-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061581
|
enQuireBio™
QP7918-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071581
|
enQuireBio™
QP7918-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081581
|
enQuireBio™
QP7918-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001288014.1 | |
Protein quaking Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9444 | |
Research Use Only | |
QKI | |
Recombinant Protein | |
E. coli | |
MVGEMETKEK PKPTPDYLMQ LMNDKKLMSS LPNFCGIFNH LERLLDEEIS RVRKDMYNDT LNGSTEKRSA ELPDAVGPIV QLQEKLYVPV KEYPDFNFVG RILGPRGLTA KQLEAETGCK IMVRGKGSMR DKKKEEQNRG KPNWEHLNED LHVLITVEDA QNRAEIKLKR AVEEVKKLLV PAAEGEDSLK KMQLMELAIL NGTYRDANIK SPALAFSLAA TAQAAPRIIT GPAPVLPPAA LRTPTPAGPT IMPLIRQIQT AVMPNGTPHP TAAIVPPGPE AGLIYTPYEY PYTLAPATSI LEYPIEPSGV LGAVATKVRR HDMRVHPYQR IVTADRAATG N | |
Greater than 80% as determined by SDS-PAGE. |