missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSMA2 Protein

A cDNA sequence encoding the PSMA2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13181-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932759

  • 120.00 € / 5µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PSMA2 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PSMA2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.1M NaCl, 1mM DTT and 0.1mM PMSF.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAERGYSFSL TTFSPSGKLV QIEYALAAVA GGAPSVGIKA ANGVVLATEK KQKSILYDER SVHKVEPITK HIGLVYSGMG PDYRVLVHRA RKLAQQYYLV YQEPIPTAQL VQRVASVMQE YTQSGGVRPF GVSLLICGWN EGRPYLFQSD PSGAYFAWKA TAMGKNYVNG KTFLEKRYNE DLELEDAIHT AILTLKESFE GQMTEDNIEV GICNEAGFRR LTPTEVKDYL AAIA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers